missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 2.0N Standardized Solution, Alfa Aesar™

Brand:  Alfa Aesar 035631.K7

Code : NEW

Additional Details : CAS Number : 1310-73-2 Weight : 0.01000kg Transport : UN number : 1824 Chem class : 1b Pack group : II

Quantity 4L

 View more versions of this product

Product Code. 11323757

  • £65.60 / 4 L


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Sodium hydroxide
Liquid
40
1310-73-2
NaOH
MFCD00003548

Specifications

Sodium hydroxide
Colorless solution
NaOH
MFCD00003548
Colorless
1310-73-2
Liquid
40
Odorless
4L
Videos
SDS