missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number 15558344
110.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Poly Bottle
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Quantity 1 L
Formula Weight 40
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.05kg
Chemical Name or Material Sodium Hydroxide, 1N Volumetric Solution
compliance-icons
Product Identifier
  • Sodium Hydroxide 1 mol/l
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'