missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 10.0N Standardized Solution, Thermo Scientific Chemicals

Catalog Number 15411357
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Sodium hydroxide, 10.0N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
CAS 1310-73-2
Molecular Formula HNaO
MDL Number MFCD00003548
UN Number UN1824
Concentration 10.0N
Density 1.327 g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether.
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium hydroxide, Standardized Solution
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'