missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 1.0N Solution, Biotech Reagent, J.T.Baker™

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Brand:  JT Baker 0328-08

Additional Details : Weight : 19.00000kg Transport : UN number : 1824 Chem class : 8 Pack group : II

Packaging Poly Drum
Quantity 19L

 View more versions of this product

Product Code. 15577814

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide

Specifications

Sodium Hydroxide, 1.0N Solution
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Poly Drum
19L
1310-73-2
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Biotech Reagent
1L = 1.05kg
Videos
Documents