missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Polyethylene Bottle
Specifications
Quantity | 1 L |
Formula Weight | 40 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Narrow Mouth Polyethylene Bottle |
Specific Gravity | 1L = 1.02kg |
Chemical Name or Material | Sodium Hydroxide, 0.5N Volumetric Solution |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'