missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
98.80 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Poly Bottle
Specifications
Quantity | 1 L |
Formula Weight | 40 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Narrow Mouth Poly Bottle |
Specific Gravity | 1L = 1.00kg |
Chemical Name or Material | Sodium Hydroxide, 0.1N Volumetric Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'