Learn More
Sodium hydrosulfite, ca. 85%, Tech., ACROS Organics™
Brand: Acros Organics 169590020
Code : 29
Additional Details : CAS Number : 7775-14-6 Weight : 2.05000kg Transport : UN number : 1384 Chem class : 1 Pack group : II
Quantity | 2kg |
---|---|
Packaging | Glass Bottle |
Description
Reduction reagentChemical Identifiers
Sodium hydrosulfite | |
7775-14-6 | |
≈85% | |
Na2O4S2 | |
174.1 | |
Sodium dithionite |
tech., ca. 85% | |
Technical Grade | |
ca. 85% | |
Na2S2O4 | |
MFCD00011640 |
Specifications
Sodium hydrosulfite | |
7775-14-6 | |
Crystalline Powder | |
ca. 85% | |
Na2S2O4 | |
MFCD00011640 | |
Sodium dithionite | |
Glass Bottle | |
> 300.0°C |
tech., ca. 85% | |
Technical Grade | |
≈85% | |
Na2O4S2 | |
174.1 | |
15, 8747 | |
Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol | |
White | |
2kg |