missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfide hydrate, pure, flakes, Thermo Scientific Chemicals

Catalog Number 10648314
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
207683-19-0
HNaS
56.06
MFCD00149796
HYHCSLBZRBJJCH-UHFFFAOYSA-M
sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp
16211346
sodium sulfanide
[Na+].[SH-]
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
207683-19-0
HNaS
56.06
MFCD00149796
HYHCSLBZRBJJCH-UHFFFAOYSA-M
sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp
16211346
sodium sulfanide
[Na+].[SH-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 55.0°C
Physical Form Deliquescing Flakes
Quantity 10 kg
Linear Formula NaSH·xH2O
Fieser 03,266
Flash Point 90°C
Merck Index 14,8587
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 56.06
Percent Purity ≥67% min. (NaHS) typically 70 to 72%
Grade Pure
Odor Obnoxious
Packaging Plastic drum
Chemical Name or Material Sodium hydrosulfide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'