missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Carbonate, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical S/4200/63

Code : 21

Additional Details : CAS Number : 144-55-8 Weight : 3.00000kg

Packaging Plastic powder jar
Quantity 3kg

 View more versions of this product

Product Code. 10010510

Quantity Price
1 £45.99 / 3kg
4 £40.03 / 3kg
16 £38.03 / 3kg
In stock, ready to ship: 44

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Sodium Hydrogen Carbonate, Extra Pure, SLR

Specifications

Chemical Identifiers

144-55-8
84.006
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium;hydrogen carbonate
CHNaO3
3528
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
C(=O)(O)[O-].[Na+]

Specifications

144-55-8
3528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
84.01g/mol
Plastic powder jar
270°C
3kg
CHNaO3
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
C(=O)(O)[O-].[Na+]
84.006
CHEBI:32139
Powder Solid
White
8.3
Videos
SDS Documents
Certificates