missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
178.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
178.03 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
178.03 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 500g, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M
Specifications
Melting Point | 270°C |
Color | White |
pH | 8.3 |
Physical Form | Powder/Solid |
Quantity | 5 kg |
IUPAC Name | sodium hydrogen carbonate |
Formula Weight | 84.01g/mol |
Packaging | Plastic keg |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'