missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, ACROS Organics™

Brand:  Acros Organics 189860050

Code : 29

Additional Details : CAS Number : 7646-69-7 Weight : 5.05000kg Transport : UN number : 1427 Chem class : 4.3 Pack group : I

Quantity 5kg

 View more versions of this product

Product Code. 10447903

  • £581.20 / 5kg


EU Stock 4
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

Adsorbent, Reduction reagent
Specifications

Chemical Identifiers

Sodium hydride, 60% dispersion in mineral oil
57 to 63% (NaH)
NaH
MFCD00003471
8042-47-5
HNa
24

Specifications

Sodium hydride, 60% dispersion in mineral oil
Tacky Powder
HNa
24
02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
800.0°C
165°C
8042-47-5
57 to 63% (NaH)
NaH
MFCD00003471
15, 8760
1.2000g/mL
Light gray to slightly yellow
5kg
Videos
SDS