missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexametaphosphate, Pure, Flake, Mainly (NaPO3)6, Fisher Chemical™

Catalog Number p-7073525
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
10124-56-8
Na6O18P6
136045
This item is not returnable. View return policy
10124-56-8
Na6O18P6
136045

Na6O18P6, CAS Number-10124-56-8, 1kg, White, 611.7708g/mol, 640 deg.C, 136045, Plastic powder jar, 6.0 to 7.7, Solid

TRUSTED_SUSTAINABILITY
Melting Point 640°C
Color White
pH 6.0 to 7.7
Physical Form Solid
Quantity 500 g
Formula Weight 611.7708g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'