missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hexametaphosphate, Pure, Flake, Mainly (NaPO3)6, Fisher Chemical™
Click to view available options
Quantity:
500 g
Description
Na6O18P6, CAS Number-10124-56-8, 1kg, White, 611.7708g/mol, 640 deg.C, 136045, Plastic powder jar, 6.0 to 7.7, Solid
Specifications
Melting Point | 640°C |
Color | White |
pH | 6.0 to 7.7 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 611.7708g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'