missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium hexaiodoplatinate(IV) hexahydrate, Premion™, 99.95% (metals basis), Pt 17.1% min

Brand:  Alfa Aesar™ 041018.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11319427

  • £53.42 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Sodium hexaiodoplatinate(IV) hexahydrate
Premion™
1110.58 (1002.50 Anhydrous)
MFCD00798546

Specifications

Sodium hexaiodoplatinate(IV) hexahydrate
Powder
(metals basis)
MFCD00798546
1g
Premion™
99.95%
1110.58 (1002.50 Anhydrous)
Pt 17.1% min.
Videos
SDS