missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium hexaiodoplatinate(IV) hexahydrate, Premion™, 99.95% (metals basis), Pt 17.1% min

Brand:  Alfa Aesar™ 041018.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11309427

  • £199.92 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Sodium hexaiodoplatinate(IV) hexahydrate
Premion™
1110.58 (1002.50 Anhydrous)
MFCD00798546

Specifications

Sodium hexaiodoplatinate(IV) hexahydrate
Powder
(metals basis)
MFCD00798546
5g
Premion™
99.95%
1110.58 (1002.50 Anhydrous)
Pt 17.1% min.
Videos
SDS