missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals

Catalog Number p-7080463
Click to view available options
Quantity:
10 g
50 g
12005-86-6
AsF6Na
211.902
MFCD00014228
NFXMAZFYHDSPPB-UHFFFAOYSA-N
sodium hexafluoroarsenate v, potassium hexafluoro-$l^ 5-arsanuide, sodium hexafluoro-??-arsanuide, sodium hexafluoroarsenate, naasf6
14205726
sodium;hexafluoroarsenic(1-)
F[As-](F)(F)(F)(F)F.[Na+]
12005-86-6
AsF6Na
211.902
MFCD00014228
NFXMAZFYHDSPPB-UHFFFAOYSA-N
sodium hexafluoroarsenate v, potassium hexafluoro-$l^ 5-arsanuide, sodium hexafluoro-??-arsanuide, sodium hexafluoroarsenate, naasf6
14205726
sodium;hexafluoroarsenic(1-)
F[As-](F)(F)(F)(F)F.[Na+]

Sodium hexafluoroarsenate(V) is generally used for pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 10 g
UN Number UN1557
Solubility Information Soluble in water.
Formula Weight 211.9
Percent Purity 99%
Chemical Name or Material Sodium hexafluoroarsenate (V)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'