missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min

Brand:  Alfa Aesar™ 011890.06

Code : NEW

Additional Details : CAS Number : 19567-78-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11321988

  • £252.20 / 5g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Sodium hexachloroiridate(IV) hexahydrate
19567-78-3
558.99 (450.92 Anhydrous)
MFCD00149169

Specifications

Sodium hexachloroiridate(IV) hexahydrate
Powder
MFCD00149169
600°C (dec.)
5g
19567-78-3
558.99 (450.92 Anhydrous)
Red-black
Ir 33.9% min.
Videos
SDS