missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexabromoosmate(IV), Premion™, 99.95% (metals basis), Os 26.1% min, Thermo Scientific™

Catalog Number p-7051082
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
MFCD00798532
This item is not returnable. View return policy
MFCD00798532

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Black to Red
Physical Form Crystalline Powder
Quantity 1 g
Linear Formula Na2OsBr6
Formula Weight 715.64
Percent Purity 99.95%
Grade Premion™
Concentration or Composition (by Analyte or Components) Os 26.1% min.
Assay (metals basis)
Chemical Name or Material Sodium hexabromoosmate(IV)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'