missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium hexabromoosmate(IV), Premion™, 99.95% (metals basis), Os 26.1% min

Brand:  Alfa Aesar™ 040412.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11314018

  • £610.40 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Sodium hexabromoosmate(IV)
Na2OsBr6
MFCD00798532
Premion™
715.64

Specifications

Sodium hexabromoosmate(IV)
Crystalline Powder
(metals basis)
715.64
Os 26.1% min.
5g
Premion™
99.95%
Na2OsBr6
MFCD00798532
Red to black
Videos
SDS