missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycolate, 97%, ACROS Organics™

Brand:  Acros Organics 351570250

Code : 29

Additional Details : CAS Number : 2836-32-0 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10694272

  • £12.20 / 25g


EU Stock 9
Add to basket

Specifications

Chemical Identifiers

Sodium glycolate
2836-32-0
97%
NaOOCCH2OH
MFCD00065358
97%
97%
C2H3NaO3
98.03
Sodium hydroxyacetate

Specifications

Sodium glycolate
2836-32-0
97%
Authentic
NaOOCCH2OH
MFCD00065358
Sodium hydroxyacetate
White to off-white
97%
Powder
97%
C2H3NaO3
98.03
Glass bottle
Solubility in water: soluble.
25g
Videos
SDS