missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycocholate hydrate, 98%, ACROS Organics™

Brand:  Acros Organics 369460250

Code : 29

Additional Details : CAS Number : 207614-05-9 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10492865

  • £1056.80 / 25g


Estimated Shipment
09-03-2018
Add to basket


Specifications

Chemical Identifiers

Sodium glycocholate hydrate
207614-05-9
98%
487.61
Glycocholic acid sodium salt
98%
≥97.5% (on dry basis) (Total base)
C26H42NNaO6
MFCD09037360

Specifications

Sodium glycocholate hydrate
207614-05-9
≥97.5% (on dry basis) (Total base)
Authentic
487.61
Glass bottle
+28° to +31° (20°C, 589nm) (c=1, H2O) on dry basis
+ 29.50 (20.00°C c=1,H2O)
25g
98%
Liquid
98%
C26H42NNaO6
MFCD09037360
Glycocholic acid sodium salt
8% max. (K.F.)
Clear colorless to slightly yellow
Videos
SDS