missing translation for 'onlineSavingsMsg'
Learn More

Sodium glucoheptonate, Thermo Scientific Chemicals

Catalog Number p-7043276
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
31138-65-5
C7H13NaO8
248.163
MFCD00013425
FMYOMWCQJXWGEN-AXUKCSBQSA-M
sodium glucoheptonate, sodium 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoate, d-sodium glucoheptonate, sodium-alpha-glucoheptonate, 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoic acid sodium salt
49853507
sodium;(2R,3R,4R,5S,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
C(C(C(C(C(C(C(=O)[O-])O)O)O)O)O)O.[Na+]
This item is not returnable. View return policy
31138-65-5
C7H13NaO8
248.163
MFCD00013425
FMYOMWCQJXWGEN-AXUKCSBQSA-M
sodium glucoheptonate, sodium 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoate, d-sodium glucoheptonate, sodium-alpha-glucoheptonate, 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoic acid sodium salt
49853507
sodium;(2R,3R,4R,5S,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
C(C(C(C(C(C(C(=O)[O-])O)O)O)O)O)O.[Na+]

Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

Solubility
Freely soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 100 g
Solubility Information Freely soluble in water
Formula Weight 248.16
Chemical Name or Material Sodium glucoheptonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'