missing translation for 'onlineSavingsMsg'
Learn More

Sodium fumarate, 98%, Thermo Scientific Chemicals

Catalog Number p-7081338
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
17013-01-3
C4H4Na2O4
162.052
MFCD00064567
VXXVUHAXJHEYFH-SEPHDYHBSA-N
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
87110070
(E)-but-2-enedioic acid;sodium
C(=CC(=O)O)C(=O)O.[Na].[Na]
This item is not returnable. View return policy
17013-01-3
C4H4Na2O4
162.052
MFCD00064567
VXXVUHAXJHEYFH-SEPHDYHBSA-N
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
87110070
(E)-but-2-enedioic acid;sodium
C(=CC(=O)O)C(=O)O.[Na].[Na]

Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Assay Percent Range 98%
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 160.04
Percent Purity 98%
Chemical Name or Material Sodium fumarate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'