missing translation for 'onlineSavingsMsg'
Learn More

Sodium formaldehydesulfoxylate dihydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-3758754
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy
6035-47-8
CH3NaO32H2O
154.11
MFCD00150598
UCWBKJOCRGQBNW-UHFFFAOYSA-M
sodium hydroxymethanesulfinate dihydrate, sodium formaldehydesulfoxylate dihydrate, methanesulfinic acid, hydroxy-, monosodium salt, dihydrate, sodium formaldehyde sulfoxylate, sodium dihydrate hydroxymethanesulfinate, sodium formaldehyde sulfoxylate nf, formaldehyde sodium sulfoxylate dihydrate, monosodium hydroxymethanesulfinate dihydrate, 79-25-4 parent, hydroxymethanesulfinic acid monosodium salt dihydrate
23666330
sodium;hydroxymethanesulfinate;dihydrate
C(O)S(=O)[O-].O.O.[Na+]
This item is not returnable. View return policy
6035-47-8
CH3NaO32H2O
154.11
MFCD00150598
UCWBKJOCRGQBNW-UHFFFAOYSA-M
sodium hydroxymethanesulfinate dihydrate, sodium formaldehydesulfoxylate dihydrate, methanesulfinic acid, hydroxy-, monosodium salt, dihydrate, sodium formaldehyde sulfoxylate, sodium dihydrate hydroxymethanesulfinate, sodium formaldehyde sulfoxylate nf, formaldehyde sodium sulfoxylate dihydrate, monosodium hydroxymethanesulfinate dihydrate, 79-25-4 parent, hydroxymethanesulfinic acid monosodium salt dihydrate
23666330
sodium;hydroxymethanesulfinate;dihydrate
C(O)S(=O)[O-].O.O.[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 63.0°C to 64.0°C
Assay Percent Range 97.5% min. (Iodometry)
Packaging Plastic bottle
Linear Formula HOCH2SO2Na·2H2O
Quantity 1 kg
Merck Index 15,8755
Solubility Information Solubility in water: 1088g/L. Other solubilities: practically insoluble in abs.alcohol, ether and, benzene
Formula Weight 154.11
Percent Purity 98%
Chemical Name or Material Sodium formaldehydesulfoxylate dihydrate
compliance-icons
Product Identifier
  • Sodium formaldehydesulfoxylate dihydrate
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H341-Suspected of causing genetic defects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'