missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fluoride, Powder, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Powder |
Quantity | 500 g |
Formula Weight | 41.99 |
Grade | USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.78kg |
Chemical Name or Material | Sodium Fluoride |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH032-Contact with acids liberates very toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'