missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407883
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
sodium;fluoride
[F-].[Na+]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
sodium;fluoride
[F-].[Na+]

It is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including its use in synthetic chemistry, for example in Finkelstein reaction for preparing fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1704°C
Melting Point 993°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 41.99
Odor Odorless
Refractive Index 1.336
Appearance White powder
Vapor Pressure 1.3 hPa (1mm Hg) at 1077°C
Density 2.78
Chemical Name or Material Sodium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'