missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 97%, extra pure, ACROS Organics™

Brand:  Acros Organics 191270250

Code : 29

Additional Details : CAS Number : 7681-49-4 Weight : 0.07500kg Transport : UN number : 1690 Chem class : 1 Pack group : III


Quantity 25g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10711281

  • £11.50 / 25g


EU Stock 13
Add to basket


Specifications

Chemical Identifiers

Sodium fluoride
7681-49-4
97%
FNa
41.98
97%
Extra Pure
97%
NaF
MFCD00003524

Specifications

Sodium fluoride
7681-49-4
Powder or Cyrstals
97%
NaF
MFCD00003524
15, 8754
100ppm max.
Plastic bottle
Solubility in water: 40g/L (25°C). Other solubilities: slightly soluble in alcohol
1700.0°C
993.0°C
97%
Extra Pure
97%
FNa
41.98
01,1073; 02,382
120ppm max.
200ppm max.
1% max. (Na2SiF6)
5000ppm max.
White
25g
Videos
SDS