missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T. Baker™
729.60 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
1 kg
2 kg
Packaging:
POLYSTORMOR
Poly Pail
Poly bottle
Description
- For Electrophoresis and Protein Solubilization; For Laboratory,Research,or Manufacturing Use
Specifications
Quantity | 1 kg |
Flash Point | 170.00°C |
IUPAC Name | sodium;dodecyl sulfate |
Formula Weight | 288.38 |
Grade | Ultrapure Bioreagent |
Packaging | Poly bottle |
Specific Gravity | 1L = 0.40kg |
Chemical Name or Material | Sodium Dodecyl Sulfate |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'