missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
25 g
100 g
1 kg
2 kg
Packaging:
POLYSTORMOR
Poly Bottle
Poly Pail
Description
- For Electrophoresis and Protein Solubilization
Specifications
Quantity | 25 g |
Flash Point | 170.00°C |
IUPAC Name | sodium dodecyl sulfate |
Formula Weight | 288.38 |
Grade | Ultrapure Bioreagent |
Packaging | POLYSTORMOR |
Specific Gravity | 1L = 0.40kg |
Chemical Name or Material | Sodium Dodecyl Sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'