missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate, PlusOne™, Cytiva
Click to view available options
Quantity:
100 g
Specifications
CAS | 151-21-3, 7732-18-5 |
Assay Percent Range | ≥98.5% (sodium dodecyl sulfate C12 compounds) |
Molecular Formula | C12H25NaO4S |
MDL Number | MFCD00036175 |
Quantity | 100 g |
Synonym | sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
IUPAC Name | sodium dodecyl sulfate |
Molecular Weight (g/mol) | 288.38 |
Show More |
Safety and Handling
Signal Word
- Warning
- Flammable solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'