missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number 11324278
Click to view available options
Quantity:
250 mL
1 L
72-17-3
C3H5NaO3
112.06
MFCD00065400
NGSFWBMYFKHRBD-UHFFFAOYSA-M
sodium lactate, lactic acid sodium salt, monosodium lactate, sodium dl-lactate, lacolin, per-glycerin, lactic acid, monosodium salt, sodium alpha-hydroxypropionate, monosodium 2-hydroxypropanoate, propanoic acid, 2-hydroxy-, monosodium salt
23666456
CHEBI:75228
sodium;2-hydroxypropanoate
CC(C(=O)[O-])O.[Na+]
72-17-3
C3H5NaO3
112.06
MFCD00065400
NGSFWBMYFKHRBD-UHFFFAOYSA-M
sodium lactate, lactic acid sodium salt, monosodium lactate, sodium dl-lactate, lacolin, per-glycerin, lactic acid, monosodium salt, sodium alpha-hydroxypropionate, monosodium 2-hydroxypropanoate, propanoic acid, 2-hydroxy-, monosodium salt
23666456
CHEBI:75228
sodium;2-hydroxypropanoate
CC(C(=O)[O-])O.[Na+]

Useful chiral synthon; building block for depsipeptides

Useful chiral synthon; building block for depsipeptides.Sodium DL-lactate, 60% w/w aq. Soln is used as a food additive, plasticizer for casein, preservative, acidity regulator, bulking agent, and in shampoo and liquid soaps. It is also used for the treatment for arrhythmias and sympathomimetics. It is useful chiral synthon and building block for depsipeptides. It is used as a corrosion inhibitor in alcohol antifreeze mixtures, flavoring agent, emulsifier, corrosion inhibitor and pH control agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.326 g/mL
Odor Odorless
Linear Formula C3H5NaO3
Quantity 250 mL
Beilstein 4332999
Merck Index 14,8635
Solubility Information Miscible with water and alcohol.
Formula Weight 112.06
Concentration or Composition (by Analyte or Components) 60% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Sodium DL-lactate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'