missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Sodium dihydrogen phosphate monohydrate is used as a food additive. It finds use as mordant in dyeing. It is also used in the manufacture of enamels, chemicals, pharmaceuticals, ceramics, detergents, and boiler compounds. In addition, it is used as a fireproofing agent, thickening agent, emulsifier, and as a reagent and buffer in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 100°C to H2O |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.998% (metals basis) |
Solubility Information | Soluble in water; insoluble in ethanol,ether and chloroform. |
Formula Weight | 137.99 (119.98 Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'