missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
10 kg
Description
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
Solubility
Soluble in water.(1 M at 20°C)
Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
Solubility
Soluble in water.(1 M at 20°C)
Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
Specifications
Quantity | 2500 g |
Linear Formula | NaH2PO4·2H2O |
Merck Index | 14,8660 |
Solubility Information | Soluble in water.(1 M at 20°C) |
SMILES | O.O.OP(=O)(O)[O-].[Na+] |
Molecular Weight (g/mol) | 156.006 |
Formula Weight | 156.01 (119.98 Anhydrous) |
Percent Purity | 99% |
Sensitivity | Hygroscopic |
Density | 1.91 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'