missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dihydrogen Orthophosphate Dihydrate, Extra Pure, SLR, Crystal, Fisher Chemical™

£42.04 - £252.00

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.006
MDL Number 149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10617042
SDS View Documents
Fisher Chemical
S/3720/53
500g Plastic powder jar £42.04
500g
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 51
Please to purchase this item. Need a web account? Register with us today!
10429380
SDS View Documents
Fisher Chemical
S/3720/60
1kg Plastic powder jar £69.82
1kg
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 26
Please to purchase this item. Need a web account? Register with us today!
10010370
SDS View Documents
Fisher Chemical
S/3720/65
5kg Plastic keg £252.00
5kg
Qty Price Break Available View alternative products
Save Up to 
»
In stock, ready to ship: 21
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

13472-35-0
156.006
VBJGJHBYWREJQD-UHFFFAOYSA-M
23673460
O.O.OP(=O)(O)[O-].[Na+]
H6NaO6P
149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
sodium;dihydrogen phosphate;dihydrate

Specifications

13472-35-0
149209
VBJGJHBYWREJQD-UHFFFAOYSA-M
sodium;dihydrogen phosphate;dihydrate
23673460
Solid
White
4.5
H6NaO6P
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
O.O.OP(=O)(O)[O-].[Na+]
156.006
156.01g/mol
Plastic powder jar
60°C
500g
SDS