missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dihydrogen Orthophosphate 0.25M Solution, Certified, HPLC for Gradient Analysis, Fisher Chemical
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Specifications
CAS | 7558-80-7 |
pH | 4.5 |
Physical Form | Liquid |
Quantity | 500 mL |
Molecular Formula | H2NaO4P |
MDL Number | MFCD00003527,MFCD00146206 |
Synonym | monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
SMILES | [Na+].OP(O)([O-])=O |
IUPAC Name | sodium;dihydrogen phosphate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'