missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Sodium deuteroxide is a reagent used in the synthesis of rac Zearalenone-d6. is used in many chemical reactions and industrial practices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 10 g |
Assay Percent Range | 99.5% |
Solubility Information | Fully miscible in water. |
Formula Weight | 41.01 |
Odor | Odorless |
Refractive Index | 1.418 |
Sensitivity | Air and moisture sensitive |
Appearance | Colorless liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'