Learn More
Thermo Scientific Alfa Aesar Sodium deuteroxide, 30% w/w solution in D2O, 99.5% (Isotopic)
Brand: Thermo Scientific Alfa Aesar 042345.09
Sodium deuteroxide, 30% w/w solution in D2O, 99.5% (Isotopic)
Code : NEW
Additional Details : CAS Number : 14014-06-3 Weight : 0.01000kg Transport : UN number : 1824 Chem class : 8 Pack group : II