missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium deuteroxide, 30% w/w solution in D2O, 99.5% (Isotopic)

NMR Solvent

Brand:  Alfa Aesar™ 042345.18

Code : NEW

Additional Details : CAS Number : 14014-06-3 Weight : 0.05000kg Transport : UN number : 1824 Chem class : 8 Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11394438

  • £116.60 / 50g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

Sodium deuteroxide
Isotopic
MFCD00037669
14014-06-3
41.01
UN1824

Specifications

Sodium deuteroxide
1.462g/mL
1.4039
50g
14014-06-3
99.5%
MFCD00037669
30% w/w solution in D2O
Odorless
Air & Moisture Sensitive
Isotopic
Liquid
41.01
UN1824
Videos
SDS