missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cyanide, Reagent Grade, 97%, Honeywell™
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Boiling Point | 1500°C |
Melting Point | 563.7°C |
Color | White |
pH | 11 to 12 (49g/L at 25°C) |
Quantity | 1 kg |
Linear Formula | NaCN |
UN Number | UN1689 |
Beilstein | 3587243 |
Solubility Information | In water 49g/L at 20°C |
Formula Weight | 49.01g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'