missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate, Dihydrate, Granular, U.S.P. - F.C.C., J.T. Baker™
302.10 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Granules |
Quantity | 2.5 kg |
Formula Weight | 294.1 |
Grade | FCC/USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.70kg |
Chemical Name or Material | Sodium Citrate, Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'