missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium chromate, anhydrous, 98%

Brand:  Alfa Aesar™ A10547.0E

Code : NEW

Additional Details : CAS Number : 7775-11-3 Weight : 2.50000kg Transport : UN number : 3288 Chem class : 6.1 Pack group : II

Quantity 2500g

 View more versions of this product

Product Code. 11418833

  • £218.40 / 2500g


EU Stock 3
Add to basket

Description

Description

Applications
Sodium chromate anhydrous is used as a corrosion inhibitor in the petroleum industry, wood preservative and dyeing auxiliary in the textile industry. It is also used to prepare pigments for paints and inks. It also finds use in pharmaceuticals to determine red blood cell volume. In synthetic chemistry, it is utilized as an oxidant to convert primary alcohols and secondary alcohols to carboxylic acids and ketones respectively.

Solubility
Soluble in water. Slightly soluble in methanol and ethanol.

Notes
Hygroscopic. Incompatible with combustible materials and reducing agents.
Specifications

Chemical Identifiers

Sodium chromate
Anhydrous
161.97
UN3288
7775-09-9
Na2CrO4
MFCD00003484
14,8601

Specifications

Sodium chromate
7775-09-9
Na2CrO4
MFCD00003484
14,8601
792°C
Anhydrous
98%
161.97
UN3288
Odorless
2500g
Videos
SDS