missing translation for 'onlineSavingsMsg'
Learn More

Sodium chromate, 98+%, anhydrous, Thermo Scientific Chemicals

Catalog Number 10774184
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7775-11-3
CrNa2O4
191.97
MFCD00003484
PXLIDIMHPNPGMH-UHFFFAOYSA-N
sodium chromate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
[Na+].[Na+].[O-][Cr]([O-])(=O)=O
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7775-11-3
CrNa2O4
191.97
MFCD00003484
PXLIDIMHPNPGMH-UHFFFAOYSA-N
sodium chromate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
[Na+].[Na+].[O-][Cr]([O-])(=O)=O

Sodium chromate, 98+%, CrNa2O4, CAS Number-7775-11-3, asex, atlacide, chlorate de sodium, chloric acid, sodium salt, chlorsaure, desolet, kusatol, rasikal, sodium chlorate, tumbleleaf, 500g, 98% min. (Iodometry), CHEBI:65242, Yellow, 231-889-5, 161.97, GHS Signal Word: Danger

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 792.0°C
Color Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula Na2CrO4
Merck Index 15, 8736
Solubility Information Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in ethanol
IUPAC Name sodium chromate
Formula Weight 161.97
Percent Purity ≥98%
Packaging Plastic bottle
Chemical Name or Material Sodium chromate, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'