missing translation for 'onlineSavingsMsg'
Learn More

STE Buffer, Sodium Chloride-Tris-EDTA, 10X Solution, pH 8.0, Fisher BioReagents

Brand:  Fisher Bioreagents BP2479-500

Code : 32

Additional Details : Weight : 0.55000kg

Quantity 500mL
Packaging Poly Bottle

 View more versions of this product

Product Code. 10274633

  • £86.80 / 500mL


Estimated Shipment
13-03-2018
Add to basket

Description

Description

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X STE buffer solution - simply dilute as needed
  • When diluted, 1X solution contains 10mM tris, 1mM EDTA, and 100mM NaCl with pH 8.0 at 25°C
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • STE buffer is used for DNA extraction protocols
  • Intended for laboratory research use only
Specifications

Chemical Identifiers

Sodium Chloride-Tris-EDTA (STE) Buffer
10X, pH 8.0
7732-18-5
STE

Specifications

Sodium Chloride-Tris-EDTA (STE) Buffer
7732-18-5
Water:>90% Sodium Chloride: <10% 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-hydrochloride: <5% Ethylenediamine tetraacetic acid (EDTA): <1.0%
STE
Protease free
100°C
Clear
10X, pH 8.0
Liquid
DNase free
Poly Bottle
Filtered through a 5-micron filter.
8
500mL
Videos
SDS Documentation