missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, Spectroscopy Grade, Thermo Scientific Chemicals

Catalog Number 15471907
Click to view available options
Quantity:
75 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1413°C
Melting Point 801°C
CAS 7647-14-5
Color White
pH 6 to 9
Physical Form Powder
Quantity 75 g
Molecular Formula ClNa
MDL Number MFCD00003477
Merck Index 14,8599
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Grade Spectroscopy
Odor Odorless
Refractive Index 1.5442
Sensitivity Hygroscopic
Density 2.165 g/mL
Chemical Name or Material Sodium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'