missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride Solution, 26% (saturated in water at 20°C, AT), Honeywell™ Fluka™
Click to view available options
Quantity:
1 L
5 L
Packaging:
Poly bottle
Specifications
Color | Colorless |
pH | 6.0 to 8.0 |
Physical Form | Liquid |
Quantity | 1 L |
Linear Formula | NaCl |
UN Number | NONH for all modes of transport |
Beilstein | 3534976 |
Formula Weight | 58.44g/mol |
Percent Purity | 26% (saturated in water at 20°C, AT) |
Packaging | Poly bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'