missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride Solution, 26% (saturated in water at 20°C, AT), Honeywell™ Fluka™

Catalog Number p-7120780
Click to view available options
Quantity:
1 L
5 L
Packaging:
Poly bottle
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
sodium chloride
[Na+].[Cl-]
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
sodium chloride
[Na+].[Cl-]
Color Colorless
pH 6.0 to 8.0
Physical Form Liquid
Quantity 1 L
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
Formula Weight 58.44g/mol
Percent Purity 26% (saturated in water at 20°C, AT)
Packaging Poly bottle
Density 1.190g/cm3
Chemical Name or Material Sodium chloride solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'