missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ACS, ≥99.5%, Honeywell™ Fluka™

Catalog Number p-8504056
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
6 x 1 kg
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
[Na+].[Cl-]
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
[Na+].[Cl-]

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1465°C
Melting Point 800°C
Color Colorless
pH 7.0 (at 20°C)
Physical Form Crystalline
Quantity 500 g
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
Solubility Information 358g/L at 20°C
IUPAC Name sodium;chloride
Formula Weight 58.44g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly bottle
Density 2.160g/cm3 at 20°C
Chemical Name or Material Sodium chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'