missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, Crystal, Ultrapure Bioreagent, J.T. Baker™
85.17 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- For Liquid Chromatography and Molecular Biology applications
Specifications
Physical Form | Crystals |
Quantity | 500 g |
Formula Weight | 58.44 |
Grade | Ultrapure Bioreagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.16kg |
Chemical Name or Material | Sodium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'