missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium chloride crystal optic rectangle, 41mm x 23mm x 6mm, unpolished

Brand:  Alfa Aesar™ 038399.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15446205

  • £28.70 / Each


Estimated Shipment date
from Supplier 06-12-2017
Add to basket

Specifications

Specifications

Sodium chloride crystal optic rectangle
41mm x 23mm x 6mm, Unpolished
1 each
Videos
SDS