missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium chloride crystal optic rectangle, 41mm x 23mm x 6mm, polished both sides

Brand:  Alfa Aesar™ 038479.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1 each

Product Code. 15416255

  • £46.96 / Each


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Specifications

Sodium chloride crystal optic rectangle
41mm x 23mm x 6mm, Polished both sides
7647-14-5
1 each
Videos
SDS